Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2V2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003350.2 | NP_003341.1 | 2, 3 | 192 | ||
XM_005251300.2 | XP_005251357.1 | 2, 3 | 251 | |||
XM_011517583.2 | XP_011515885.1 | 2, 3 | 270 | |||
XM_017013808.1 | XP_016869297.1 | 2, 3 | 177 | |||
GenBank mRNA | AF049140.1 | - | 2, 3 | 192 | ||
AK094617.1 | - | 1, 2 | 1441 | |||
AK313447.1 | - | 2, 3 | 195 | |||
BC007051.1 | - | 2, 3 | 197 | 3683051 | ||
BC016332.1 | - | 2, 3 | 217 | 4109228 | ||
BC016710.1 | - | 2, 3 | 201 | 4108684 | ||
BC028673.1 | - | 2, 3 | 194 | 4104992 | ||
BC062418.1 | - | 2, 3 | 191 | 6503261 | ||
BT006744.1 | - | 2, 3 | 171 | |||
CR407628.1 | - | 2, 3 | 171 | |||
U62136.2 | - | 2, 3 | 193 | |||
X98091.1 | - | 2, 3 | 217 |