Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLB1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286423.1 | NP_001273352.1 | 10 | 1191 | ||
NM_001286427.1 | NP_001273356.1 | 7 | 708 | |||
NM_024506.4 | NP_078782.3 | 10 | 1179 | |||
XM_011511813.2 | XP_011510115.1 | 8 | 768 | |||
XM_017004895.1 | XP_016860384.1 | 8 | 875 | |||
GenBank mRNA | AK055098.1 | - | 5 | 366 | ||
AK074393.1 | - | 4 | 359 | |||
AK299404.1 | - | 9 | 940 | |||
AK300215.1 | - | 7 | 691 | |||
AY358639.1 | - | 10 | 1159 |