Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030775.2 | NP_110402.2 | 2, 3 | 230 | ||
NM_032642.2 | NP_116031.1 | 2, 3 | 268 | |||
GenBank mRNA | AB060966.1 | - | 1, 2 | 90 | ||
AB209228.1 | - | 1, 2 | 628 | |||
AK290430.1 | - | 2, 3 | 329 | |||
BC001749.1 | - | 2, 3 | 268 | 3528819 |