Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC10A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142391.2 | NP_001135863.1 | 3 | 511 | ||
NM_001142392.2 | NP_001135864.1 | 3 | 511 | |||
NM_019848.4 | NP_062822.1 | 2 | 746 | |||
XM_005277913.4 | XP_005277970.1 | 2 | 1168 | |||
XM_006724847.3 | XP_006724910.1 | 2 | 689 | |||
XM_006724848.3 | XP_006724911.1 | 2 | 689 | |||
XM_011531200.2 | XP_011529502.2 | 2 | 721 | |||
XM_011531201.2 | XP_011529503.1 | 2 | 425 | |||
GenBank mRNA | BC004966.1 | - | 3 | 435 | 3503242 | |
BP281642.1 | - | 3 | 450 | |||
BX401493.2 | - | 3 | 483 |