Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014909.4 | NP_055724.1 | 4, 5 | 1464 | ||
XM_017021087.1 | XP_016876576.1 | 6, 7 | 539 | |||
XM_017021088.1 | XP_016876577.1 | 5, 6 | 496 | |||
XM_017021089.1 | XP_016876578.1 | 5, 6 | 355 | |||
XM_017021090.1 | XP_016876579.1 | 4, 5 | 288 | |||
GenBank mRNA | AB028959.1 | - | 4, 5 | 919 | ||
AK291377.1 | - | 4, 5 | 614 | |||
AL832588.2 | - | 4, 5 | 1168 | |||
BC051896.1 | - | 4, 5 | 758 | 5170122 |