Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPFIA2 SIRNA Assays ›
Gene Symbol:
PPFIA2
Gene Name:
PTPRF interacting protein alpha 2
Gene Aliases:
-
Chromosome Location:
Chr.12: 81257975 - 81759748 on Build GRCh38
UniGene ID:
Hs.506216
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s224985
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPFIA2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001220473.2
NP_001207402.1
26
3348
NM_001220474.2
NP_001207403.1
26
3303
NM_001220475.2
NP_001207404.1
25
3097
NM_001220476.2
NP_001207405.1
26
3142
NM_001220477.2
NP_001207406.1
23
3086
NM_001220478.2
NP_001207407.1
23
3074
NM_001220479.2
NP_001207408.1
15
1816
NM_001220480.2
NP_001207409.1
16
1749
NM_001282536.1
NP_001269465.1
24
2855
NM_003625.4
NP_003616.2
27
3211
XM_006719670.3
XP_006719733.1
25
10412
XM_011538906.2
XP_011537208.1
23
3177
XM_011538907.2
XP_011537209.1
25
3270
XM_017020079.1
XP_016875568.1
28
3204
XM_017020080.1
XP_016875569.1
26
3342
XM_017020081.1
XP_016875570.1
27
3333
XM_017020082.1
XP_016875571.1
27
3177
XM_017020083.1
XP_016875572.1
26
3321
XM_017020084.1
XP_016875573.1
27
3351
XM_017020085.1
XP_016875574.1
25
3312
XM_017020086.1
XP_016875575.1
27
3195
XM_017020087.1
XP_016875576.1
27
3320
XM_017020088.1
XP_016875577.1
27
3584
XM_017020089.1
XP_016875578.1
26
3297
XM_017020090.1
XP_016875579.1
27
3177
XM_017020091.1
XP_016875580.1
26
3321
XM_017020092.1
XP_016875581.1
26
3288
XM_017020093.1
XP_016875582.1
25
3312
XM_017020094.1
XP_016875583.1
27
3129
XM_017020095.1
XP_016875584.1
26
3270
XM_017020096.1
XP_016875585.1
25
3267
XM_017020097.1
XP_016875586.1
26
3114
XM_017020098.1
XP_016875587.1
26
3111
XM_017020099.1
XP_016875588.1
25
3288
XM_017020100.1
XP_016875589.1
26
3288
XM_017020101.1
XP_016875590.1
25
3102
XM_017020102.1
XP_016875591.1
25
3236
XM_017020103.1
XP_016875592.1
25
3279
XM_017020104.1
XP_016875593.1
25
3240
XM_017020105.1
XP_016875594.1
25
3267
XM_017020106.1
XP_016875595.1
24
3231
XM_017020107.1
XP_016875596.1
26
3114
XM_017020108.1
XP_016875597.1
26
3111
XM_017020109.1
XP_016875598.1
24
3249
XM_017020110.1
XP_016875599.1
25
3500
XM_017020111.1
XP_016875600.1
24
3231
XM_017020112.1
XP_016875601.1
24
3167
XM_017020113.1
XP_016875602.1
24
3149
XM_017020114.1
XP_016875603.1
25
3176
XM_017020115.1
XP_016875604.1
24
3167
XM_017020116.1
XP_016875605.1
24
3149
XM_017020117.1
XP_016875606.1
24
2911
XM_017020118.1
XP_016875607.1
26
3151
XM_017020119.1
XP_016875608.1
23
3104
XM_017020120.1
XP_016875609.1
25
2864
GenBank mRNA
AB210009.1
-
26
3346
AF034799.1
-
27
3219
AK123372.1
-
16
1684
AK126971.1
-
26
3263
AK294505.1
-
23
2884
AK296380.1
-
15
1787
AK299853.1
-
23
3035
AK302658.1
-
24
2826
BC104912.1
-
25
3081
8143915
BC143485.1
-
26
3126
9051993
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell-matrix adhesion
neurotransmitter secretion
glutamate secretion
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0