Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YBX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145426.1 | NP_001138898.1 | 6 | 832 | ||
NM_003651.4 | NP_003642.3 | 7 | 1039 | |||
XM_017020122.1 | XP_016875611.1 | 7 | 1133 | |||
XM_017020123.1 | XP_016875612.1 | 6 | 926 | |||
GenBank mRNA | AB209896.1 | - | 6 | 3693 | ||
AK314846.1 | - | 7 | 1008 | |||
BC008801.2 | - | 7 | 997 | 3634981 | ||
BC009744.2 | - | 6 | 785 | 3835480 | ||
BC015564.1 | - | 7 | 988 | 4137643 | ||
BC015913.1 | - | 6 | 774 | 4563632 | ||
BC021926.1 | - | 7 | 990 | 4551107 | ||
BC150580.1 | - | 2 | 157 | |||
M24069.1 | - | 7 | 1006 | |||
X72712.1 | - | 7 | 1017 | |||
X95325.1 | - | 7 | 979 |