Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271999.1 | NP_001258928.1 | 40 | 5171 | ||
NM_001272000.1 | NP_001258929.1 | 39 | 5078 | |||
NM_001272001.1 | NP_001258930.1 | 39 | 5078 | |||
NM_033407.3 | NP_212132.2 | 39 | 5105 | |||
XM_005271292.2 | XP_005271349.1 | 40 | 5161 | |||
XM_011542326.2 | XP_011540628.1 | 40 | 5188 | |||
XM_011542327.2 | XP_011540629.1 | 40 | 5188 | |||
XM_011542328.2 | XP_011540630.1 | 40 | 5188 | |||
XM_011542330.2 | XP_011540632.1 | 40 | 5188 | |||
XM_017002639.1 | XP_016858128.1 | 39 | 5095 | |||
XM_017002640.1 | XP_016858129.1 | 40 | 5188 | |||
GenBank mRNA | AB051558.1 | - | 20 | 2581 | ||
AK022097.1 | - | 1 | 901 | |||
AK055493.1 | - | 6 | 722 | |||
AK055905.1 | - | 12 | 1604 | |||
AK091479.1 | - | 13 | 1572 | |||
AK125049.1 | - | 25 | 3250 | |||
AK299737.1 | - | 11 | 1288 | |||
BC016392.1 | - | 9 | 1075 | |||
DQ118679.1 | - | 36 | 4650 | |||
DQ118680.1 | - | 36 | 4650 | |||
DQ341187.1 | - | 40 | 5078 |