Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003842.4 | NP_003833.4 | 5 | 796 | ||
NM_147187.2 | NP_671716.2 | 5 | 796 | |||
NR_027140.1 | 4 | 690 | ||||
XM_017013944.1 | XP_016869433.1 | 5 | 584 | |||
GenBank mRNA | AB451272.1 | - | 5 | 503 | ||
AB451399.1 | - | 5 | 503 | |||
AF012535.1 | - | 5 | 642 | |||
AF012628.1 | - | 5 | 503 | |||
AF016266.1 | - | 5 | 620 | |||
AF016268.1 | - | 5 | 612 | |||
AF016849.1 | - | 5 | 647 | |||
AF018657.1 | - | 5 | 503 | |||
AF018658.1 | - | 5 | 503 | |||
AF020501.1 | - | 5 | 503 | |||
AF022386.1 | - | 5 | 503 | |||
AF153687.1 | - | 5 | 621 | |||
AF192548.1 | - | 5 | 603 | |||
AK291216.1 | - | 5 | 623 | |||
AK298602.1 | - | 4 | 534 | |||
AL529460.3 | - | 5 | 796 | |||
AY358277.1 | - | 5 | 642 | |||
BC001281.1 | - | 5 | 650 | 3458466 | ||
BX538104.1 | - | 5 | 625 | |||
CR541898.1 | - | 5 | 503 | |||
FJ200482.1 | - | 4 | 397 |