Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271811.1 | NP_001258740.1 | 2 | 180 | ||
NM_004264.4 | NP_004255.2 | 2 | 180 | |||
NR_073464.1 | 2 | 430 | ||||
GenBank mRNA | AK311838.1 | - | 2 | 124 | ||
BC008380.1 | - | 2 | 152 | 4041267 | ||
CR456999.1 | - | 2 | 122 | |||
DA420057.1 | - | 2 | 391 | |||
DB024509.1 | - | 2 | 180 | |||
HY075642.1 | - | 2 | 154 | |||
KU178474.1 | - | 2 | 122 | |||
KU178475.1 | - | 2 | 122 | |||
U46837.1 | - | 2 | 131 | |||
U52960.1 | - | 2 | 127 |