Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ONECUT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004852.2 | NP_004843.2 | 1 | 1097 | ||
XM_017026096.1 | XP_016881585.1 | 1 | 1362 | |||
GenBank mRNA | BC108925.1 | - | 2 | 922 | ||
BC108926.1 | - | 2 | 922 | |||
Y18198.1 | - | 1 | 1096 |