Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164189.1 | NP_001157661.1 | 8 | 2155 | ||
NM_001164194.1 | NP_001157666.1 | 8 | 2122 | |||
NM_005817.4 | NP_005808.3 | 8 | 2158 | |||
GenBank mRNA | BC019278.1 | - | 8 | 2047 | 2905275 |