Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040662.1 | 2 | 531 | |||
GenBank mRNA | AK093953.1 | - | 3 | 445 | ||
AK290875.1 | - | 2 | 526 | |||
BC106759.2 | - | 2 | 477 | 40024782 | ||
BC113853.1 | - | 1 | 568 | |||
BC114462.1 | - | 2 | 477 | |||
BX640979.1 | - | 2 | 529 | |||
DA501560.1 | - | 2 | 532 | |||
L06175.1 | - | 2 | 527 |