Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006813.2 | NP_006804.1 | 2 | 861 | ||
GenBank mRNA | AK130046.1 | - | 2 | 819 | ||
AK130722.1 | - | 1 | 901 | |||
AK312665.1 | - | 2 | 820 | |||
AY303779.1 | - | 2 | 439 | |||
BC018112.1 | - | 2 | 731 | 3916954 | ||
BC044919.1 | - | 2 | 1041 | |||
U03105.1 | - | 2 | 789 |