Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007002.3 | NP_008933.2 | 3 | 464 | ||
NM_175573.2 | NP_783163.1 | 3 | 460 | |||
XM_005260257.1 | XP_005260314.1 | 4 | 551 | |||
XM_011528503.1 | XP_011526805.1 | 3 | 1299 | |||
XM_017027602.1 | XP_016883091.1 | 3 | 1299 | |||
XM_017027603.1 | XP_016883092.1 | 4 | 551 | |||
XM_017027604.1 | XP_016883093.1 | 3 | 1295 | |||
XM_017027605.1 | XP_016883094.1 | 3 | 1299 | |||
GenBank mRNA | AK297567.1 | - | 3 | 358 | ||
AK307459.1 | - | 394 | ||||
AL558522.3 | - | 3 | 396 | |||
BC017245.2 | - | 3 | 360 | 4890786 | ||
CR749213.1 | - | 1 | 1081 | |||
D64154.1 | - | 3 | 355 |