Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270766.1 | NP_001257695.1 | 8 | 742 | ||
NM_001270767.1 | NP_001257696.1 | 7 | 676 | |||
NM_173474.3 | NP_775745.1 | 9 | 845 | |||
XM_011522355.2 | XP_011520657.1 | 6 | 680 | |||
GenBank mRNA | AF092440.1 | - | 9 | 752 | ||
AK092915.1 | - | 5 | 2028 | |||
BC017336.2 | - | 9 | 742 | 4872717 |