Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144643.3 | NP_653244.2 | 18 | 2190 | ||
XM_017007716.1 | XP_016863205.1 | 18 | 2202 | |||
XM_017007717.1 | XP_016863206.1 | 18 | 2202 | |||
XM_017007718.1 | XP_016863207.1 | 18 | 2202 | |||
XM_017007719.1 | XP_016863208.1 | 18 | 1723 | |||
XM_017007720.1 | XP_016863209.1 | 19 | 2102 | |||
XM_017007721.1 | XP_016863210.1 | 12 | 1304 | |||
GenBank mRNA | AK055217.1 | - | 18 | 2190 | ||
AK093361.1 | - | 12 | 1343 | |||
AK122852.1 | - | 8 | 2815 |