Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM53A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013622.3 | NP_001013644.1 | 5 | 2097 | ||
NM_001174070.2 | NP_001167541.1 | 5 | 2144 | |||
NM_001297435.1 | NP_001284364.1 | 6 | 2651 | |||
XM_005247948.3 | XP_005248005.1 | 5 | 2794 | |||
XM_011513402.2 | XP_011511704.1 | 5 | 2800 | |||
XM_017007802.1 | XP_016863291.1 | 6 | 2948 | |||
GenBank mRNA | AK125566.1 | - | 2221 | |||
BC070112.1 | - | 2221 | 5268061 |