Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306149.1 | NP_001293078.1 | 1, 2 | 339 | ||
NM_001306150.1 | NP_001293079.1 | 1, 2 | 339 | |||
NM_178569.3 | NP_848664.1 | 1, 2 | 339 | |||
XM_005248256.3 | XP_005248313.1 | 1, 2 | 530 | |||
XM_005248257.3 | XP_005248314.1 | 1, 2 | 530 | |||
XM_011513975.2 | XP_011512277.1 | 1, 2 | 530 | |||
XM_011513977.2 | XP_011512279.1 | 1, 2 | 530 | |||
GenBank mRNA | AY249325.1 | - | 1, 2 | 322 | ||
BC101608.1 | - | 1, 2 | 242 | 8069114 | ||
BC101634.1 | - | 1, 2 | 242 | 8069140 | ||
BC143639.1 | - | 1, 2 | 242 | 9052153 | ||
CX754076.1 | - | 1, 2 | 224 |