Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIGD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138820.2 | NP_620175.1 | 2 | 372 | ||
GenBank mRNA | BC007502.2 | - | 2 | 372 | 2959830 |