Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175061.3 | NP_778231.2 | 2 | 370 | ||
XM_006715656.1 | XP_006715719.1 | 2 | 184 | |||
XM_017011831.1 | XP_016867320.1 | 2 | 138 | |||
XM_017011832.1 | XP_016867321.1 | 2 | 70 | |||
GenBank mRNA | AK091311.1 | - | 2 | 295 | ||
AL834234.1 | - | 1 | 89 | |||
AY372319.1 | - | 2 | 148 | |||
BC042441.1 | - | 2 | 158 | 4831063 | ||
BC047229.1 | - | 2 | 157 | |||
DA864624.1 | - | 2 | 370 |