Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MON1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286639.1 | NP_001273568.1 | 5 | 1585 | ||
NM_001286640.1 | NP_001273569.1 | 4 | 1427 | |||
NM_014940.3 | NP_055755.1 | 6 | 1912 | |||
GenBank mRNA | AB020679.1 | - | 6 | 1596 | ||
AF442486.1 | - | 5 | 1576 | |||
AK023374.1 | - | 5 | 853 | |||
AK023896.1 | - | 6 | 1640 | |||
AK128411.1 | - | 3 | 1454 | |||
AK293392.1 | - | 4 | 1147 | |||
AK296467.1 | - | 5 | 1312 | |||
BC024277.1 | - | 6 | 1892 | 4843859 |