Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012477.3 | NP_036609.1 | 3 | 527 | ||
GenBank mRNA | AF087888.1 | - | 3 | 327 | ||
AK094297.1 | - | 3 | 1359 | |||
AK315745.1 | - | 3 | 469 | |||
BC010012.2 | - | 3 | 477 | 4309279 | ||
BC064482.1 | - | 3 | 334 | 5423387 | ||
BC071626.1 | - | 3 | 476 | 3902484 | ||
BC071850.1 | - | 3 | 449 | 6425189 | ||
U79457.1 | - | 1 | 421 |
Gene Symbol: INO80B-WBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037849.1 | 7 | 1416 |