Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM19A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004440.1 | NP_001004440.1 | 5 | 1018 | ||
NM_182759.2 | NP_877436.1 | 5 | 950 | |||
XM_005270766.3 | XP_005270823.1 | 6 | 1237 | |||
XM_006710582.3 | XP_006710645.1 | 6 | 1305 | |||
GenBank mRNA | AY325116.1 | - | 5 | 950 | ||
AY325119.1 | - | 5 | 1018 |