Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304382.1 | NP_001291311.1 | 6 | 557 | ||
NM_002100.5 | NP_002091.3 | 5 | 477 | |||
XM_011531905.2 | XP_011530207.1 | 6 | 764 | |||
XM_017008137.1 | XP_016863626.1 | 5 | 2811 | |||
GenBank mRNA | BC121077.1 | - | 5 | 453 | 40119713 | |
BC121078.1 | - | 5 | 453 | 40119716 | ||
J02982.1 | - | 5 | 448 | |||
U05255.1 | - | 4 | 324 | |||
X08055.1 | - | 7 | 555 |