Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARSG SIRNA Assays ›
Gene Symbol:
ARSG
Gene Name:
arylsulfatase G
Gene Aliases:
-
Chromosome Location:
Chr.17: 68259089 - 68452000 on Build GRCh38
UniGene ID:
Hs.437249
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22626
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARSG
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001267727.1
NP_001254656.1
6
1296
NM_014960.4
NP_055775.2
6
1467
XM_005257170.3
XP_005257227.1
6
1480
XM_005257172.3
XP_005257229.1
6
1529
XM_006721777.3
XP_006721840.2
6
1494
XM_006721779.3
XP_006721842.1
6
1521
XM_011524535.2
XP_011522837.1
6
1492
XM_011524536.2
XP_011522838.1
6
1322
XM_011524537.1
XP_011522839.1
6
1243
XM_011524538.2
XP_011522840.1
6
1494
XM_011524540.2
XP_011522842.1
6
1517
XM_011524541.2
XP_011522843.1
6
1560
XM_011524542.2
XP_011522844.1
6
1560
XM_011524543.2
XP_011522845.1
6
1560
XM_011524544.2
XP_011522846.1
6
1560
XM_011524545.2
XP_011522847.1
6
1518
XM_017024360.1
XP_016879849.1
6
1332
XM_017024361.1
XP_016879850.1
6
1480
XM_017024362.1
XP_016879851.1
6
1419
XM_017024363.1
XP_016879852.1
6
1325
XM_017024364.1
XP_016879853.1
6
1321
XM_017024365.1
XP_016879854.1
6
1309
XM_017024366.1
XP_016879855.1
6
1520
XM_017024367.1
XP_016879856.1
6
1322
XM_017024368.1
XP_016879857.1
6
1322
GenBank mRNA
AB023218.1
-
5
1129
AY358380.1
-
6
1467
BC012375.1
-
5
710
3882163
Back To Top
More Information
Panther Classification:
Molecular Function -
hydrolase
Gene Ontology Categories:
Function(s)
Process(es)
glycosphingolipid metabolic process
sulfur compound metabolic process
post-translational protein modification
arylsulfatase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0