Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUFY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037442.3 | NP_001032519.1 | 6 | 1469 | ||
NM_001130709.1 | NP_001124181.1 | 6 | 917 | |||
NM_001291993.1 | NP_001278922.1 | 7 | 861 | |||
NM_001291994.1 | NP_001278923.1 | 7 | 1523 | |||
NM_014961.4 | NP_055776.1 | 6 | 1469 | |||
XM_005265656.2 | XP_005265713.1 | 7 | 1507 | |||
XM_005265658.1 | XP_005265715.1 | 6 | 783 | |||
XM_011531750.2 | XP_011530052.1 | 7 | 1105 | |||
XM_011531751.2 | XP_011530053.1 | 6 | 1118 | |||
XM_011531752.2 | XP_011530054.1 | 6 | 1118 | |||
XM_011531753.2 | XP_011530055.1 | 6 | 1118 | |||
XM_011531754.2 | XP_011530056.1 | 6 | 1118 | |||
XM_011531755.2 | XP_011530057.1 | 6 | 1083 | |||
XM_011531756.2 | XP_011530058.1 | 6 | 1083 | |||
XM_017007890.1 | XP_016863379.1 | 6 | 1118 | |||
XM_017007891.1 | XP_016863380.1 | 7 | 815 | |||
XM_017007892.1 | XP_016863381.1 | 6 | 783 | |||
GenBank mRNA | AB020678.1 | - | 6 | 1275 | ||
AF112221.1 | - | 6 | 699 | |||
AK000911.1 | - | 6 | 723 | |||
AK294421.1 | - | 6 | 654 | |||
AK296501.1 | - | 7 | 710 | |||
AK302637.1 | - | 7 | 737 | |||
BC051716.1 | - | 6 | 1337 | 5265230 | ||
BC053837.1 | - | 6 | 1299 | |||
BC071988.1 | - | 6 | 1272 | |||
BC098433.1 | - | 6 | 1299 |