Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010888.3 | NP_001010888.3 | 3 | 1007 | ||
XM_011530939.2 | XP_011529241.1 | 4 | 1252 | |||
XM_017029478.1 | XP_016884967.1 | 9 | 2518 | |||
XM_017029479.1 | XP_016884968.1 | 11 | 5811 | |||
XM_017029480.1 | XP_016884969.1 | 9 | 2552 | |||
XM_017029481.1 | XP_016884970.1 | 9 | 2547 | |||
XM_017029482.1 | XP_016884971.1 | 8 | 2457 | |||
XM_017029483.1 | XP_016884972.1 | 7 | 2253 | |||
XM_017029484.1 | XP_016884973.1 | 7 | 1885 | |||
GenBank mRNA | BC140767.1 | - | 3 | 1007 | 9021635 | |
BC171781.1 | - | 3 | 1007 | 9054435 | ||
BX647241.1 | - | 5 | 757 |