Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282550.1 | NP_001269479.1 | 5 | 1342 | ||
NM_001282551.1 | NP_001269480.1 | 5 | 1438 | |||
NM_001282552.2 | NP_001269481.1 | 4 | 1432 | |||
NM_001282554.1 | NP_001269483.1 | 4 | 1347 | |||
NM_014962.3 | NP_055777.1 | 4 | 1760 | |||
NM_181443.3 | NP_852108.2 | 5 | 1523 | |||
XM_017027728.1 | XP_016883217.1 | 6 | 1671 | |||
XM_017027729.1 | XP_016883218.1 | 6 | 1603 | |||
XM_017027730.1 | XP_016883219.1 | 3 | 1691 | |||
GenBank mRNA | AB023169.1 | - | 4 | 1708 | ||
AK296364.1 | - | 3 | 986 | |||
AK299281.1 | - | 5 | 1523 | |||
AK300371.1 | - | 3 | 1043 | |||
AK307472.1 | - | 4 | 1090 | |||
AK316256.1 | - | 4 | 1323 | |||
AK316519.1 | - | 3 | 1266 | |||
AL137756.1 | - | 1 | 2512 | |||
BC093677.1 | - | 4 | 1231 | |||
BC101527.1 | - | 4 | 1230 | 8069033 | ||
BC109315.1 | - | 5 | 1371 | 40036410 | ||
BC109316.1 | - | 5 | 1371 | 40036411 |