Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKDD1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182703.5 | NP_874362.3 | 6 | 588 | ||
GenBank mRNA | AK075298.1 | - | 4 | 339 | ||
AK098736.1 | - | 8 | 844 | |||
AK307698.1 | - | 6 | 504 | |||
BC101273.1 | - | 3 | 286 | 40024165 | ||
BC101274.1 | - | 3 | 286 | 40024168 | ||
BC101275.1 | - | 3 | 286 | 40024170 | ||
BC101276.1 | - | 3 | 279 | |||
BC110610.1 | - | 6 | 570 | |||
DR004550.1 | - | 6 | 568 |