Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102664.1 | NP_001096134.1 | 4 | 395 | ||
NM_014964.4 | NP_055779.2 | 7 | 1454 | |||
NM_148921.3 | NP_683723.2 | 6 | 1283 | |||
GenBank mRNA | AB028988.1 | - | 7 | 1419 | ||
AF062084.1 | - | 4 | 835 | |||
AF062085.1 | - | 5 | 1009 | |||
AK001491.1 | - | 5 | 1147 | |||
AK001996.1 | - | 4 | 1002 | |||
AK024115.1 | - | 6 | 1263 | |||
AK092366.1 | - | 4 | 376 | |||
AK295652.1 | - | 4 | 335 | |||
AK307455.1 | - | 4 | 336 | |||
BC070036.1 | - | 1248 | ||||
BC093972.1 | - | 4 | 908 | 7939817 | ||
BC093974.1 | - | 4 | 908 | 7939819 | ||
BC143264.1 | - | 4 | 888 | 9051766 | ||
DA220261.1 | - | 4 | 376 |