Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012507.3 | NP_001012525.1 | 1 | 598 | ||
NM_001286524.1 | NP_001273453.1 | 1 | 598 | |||
NM_001286525.1 | NP_001273454.1 | 1 | 598 | |||
NR_104462.1 | 1 | 598 | ||||
XM_017010845.1 | XP_016866334.1 | 1 | 691 | |||
GenBank mRNA | AL832235.1 | - | 1 | 598 | ||
AY902475.1 | - | 1 | 267 | |||
BC017928.1 | - | 1 | 215 | 4655418 | ||
BC039556.2 | - | 1 | 280 | 4452583 | ||
BC046178.2 | - | 1 | 230 | 4448513 | ||
BC062798.1 | - | 1 | 243 | 6495746 | ||
BM721156.1 | - | 1 | 217 | |||
BM874602.1 | - | 1 | 199 |