Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SACM1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319072.1 | NP_001306001.1 | 16 | 1529 | ||
NM_001319073.1 | NP_001306002.1 | 18 | 1697 | |||
NM_014016.4 | NP_054735.3 | 17 | 1657 | |||
XM_011533500.2 | XP_011531802.1 | 17 | 1448 | |||
GenBank mRNA | AB020658.1 | - | 17 | 1501 | ||
AK126434.1 | - | 4 | 972 | |||
AK291142.1 | - | 17 | 1541 | |||
AK296421.1 | - | 16 | 1311 | |||
AK301248.1 | - | 18 | 1537 | |||
AL136831.1 | - | 17 | 1447 | |||
BC016559.1 | - | 17 | 1453 | 3049075 | ||
CR749564.1 | - | 17 | 2593 |