Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPN1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080487.2 | NP_001073956.2 | 4 | 589 | ||
NM_001294328.1 | NP_001281257.1 | 4 | 589 | |||
XM_017023230.1 | XP_016878719.1 | 6 | 1844 | |||
GenBank mRNA | BC130001.1 | - | 4 | 589 | 40133435 | |
EU190484.1 | - | 4 | 547 |