Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003799.1 | NP_001003799.1 | 2 | 182 | ||
NM_001003806.1 | NP_001003806.1 | 2 | 182 | |||
XM_011515407.2 | XP_011513709.1 | 3 | 402 | |||
XM_017012227.1 | XP_016867716.1 | 2 | 392 | |||
XM_017012229.1 | XP_016867718.1 | 2 | 604 | |||
XM_017012230.1 | XP_016867719.1 | 2 | 208 | |||
GenBank mRNA | AF151103.1 | - | 2 | 182 | ||
BC030554.1 | - | 4 | 866 | |||
BC105589.1 | - | 3 | 400 | 4272944 | ||
BF674593.1 | - | 2 | 172 | |||
EU395807.1 | - | 2 | 161 | |||
JQ956403.1 | - | 3 | 324 | |||
KC170727.1 | - | 5 | 539 | |||
M17325.1 | - | 1 | 101 | |||
M27334.1 | - | 1 | 122 | |||
X72500.1 | - | 4 | 548 | |||
X74774.1 | - | 3 | 648 | |||
X74775.1 | - | 2 | 482 |
Gene Symbol: TRGV9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC062761.1 | - | 2 | 177 | ||
BC072387.1 | - | 3 | 499 | |||
BC072396.1 | - | 5 | 925 | |||
M16768.1 | - | 4 | 685 |