Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTFDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283035.1 | NP_001269964.1 | 4 | 385 | ||
NM_001283036.1 | NP_001269965.1 | 3 | 295 | |||
NM_001283037.1 | NP_001269966.1 | 3 | 295 | |||
NM_016407.4 | NP_057491.2 | 3 | 295 | |||
XM_017027872.1 | XP_016883361.1 | 6 | 488 | |||
GenBank mRNA | AF117231.1 | - | 3 | 243 | ||
AF161513.1 | - | 3 | 252 | |||
AF161518.1 | - | 3 | 221 | |||
AF212244.1 | - | 3 | 232 | |||
AK000481.1 | - | 3 | 225 | |||
AK000586.1 | - | 2 | 115 | |||
AK026118.1 | - | 3 | 223 | |||
AK225135.1 | - | 3 | 213 | |||
AK293662.1 | - | 3 | 223 | |||
AK302029.1 | - | 4 | 328 | |||
AK314563.1 | - | 3 | 250 | |||
BC003359.1 | - | 3 | 222 | 2900332 | ||
BG468509.1 | - | 3 | 256 | |||
BP307634.1 | - | 3 | 289 | |||
BU537939.1 | - | 3 | 205 | |||
CR983692.1 | - | 3 | 206 | |||
DC397043.1 | - | 4 | 385 |