Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PENK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135690.2 | NP_001129162.1 | 4 | 691 | ||
GenBank mRNA | AK314908.1 | - | 4 | 632 | ||
BC032505.1 | - | 2 | 542 | 5244387 | ||
BC107706.1 | - | 4 | 551 | |||
BI913625.1 | - | 2 | 546 | |||
CR541808.1 | - | 2 | 466 | |||
CR541828.1 | - | 2 | 466 |