Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C21orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286463.1 | NP_001273392.1 | 7 | 1513 | ||
NM_001286476.1 | NP_001273405.1 | 9 | 2492 | |||
NM_001286477.1 | NP_001273406.1 | 8 | 2154 | |||
NM_058180.4 | NP_478060.2 | 8 | 2526 | |||
XM_005261148.4 | XP_005261205.1 | 9 | 2229 | |||
XM_011529621.2 | XP_011527923.1 | 8 | 2509 | |||
XM_011529623.2 | XP_011527925.1 | 8 | 4514 | |||
XM_011529624.2 | XP_011527926.1 | 8 | 1389 | |||
XM_011529625.2 | XP_011527927.1 | 8 | 1663 | |||
XM_011529626.2 | XP_011527928.1 | 9 | 4860 | |||
XM_011529627.2 | XP_011527929.1 | 9 | 4940 | |||
XM_017028388.1 | XP_016883877.1 | 9 | 4691 | |||
XM_017028389.1 | XP_016883878.1 | 10 | 4577 | |||
XM_017028390.1 | XP_016883879.1 | 9 | 2858 | |||
XM_017028391.1 | XP_016883880.1 | 8 | 2681 | |||
XM_017028392.1 | XP_016883881.1 | 10 | 1878 | |||
XM_017028393.1 | XP_016883882.1 | 9 | 1677 | |||
GenBank mRNA | AK056387.1 | - | 8 | 2499 | ||
AK096641.1 | - | 7 | 2430 | |||
AK098098.1 | - | 7 | 1450 | |||
BC026042.1 | - | 9 | 2467 | |||
BM475055.1 | - | 1 | 266 |