Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIFAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204514.1 | NP_001191443.1 | 7 | 1012 | ||
NM_001204516.1 | NP_001191445.1 | 5 | 716 | |||
NM_001204517.1 | NP_001191446.1 | 6 | 1957 | |||
NM_014970.3 | NP_055785.2 | 6 | 848 | |||
XM_005244970.1 | XP_005245027.1 | 6 | 851 | |||
XM_011509307.2 | XP_011507609.2 | 6 | 851 | |||
GenBank mRNA | AK026471.1 | - | 6 | 627 | ||
AK122897.1 | - | 6 | 1959 | |||
AK297368.1 | - | 5 | 643 | |||
AK303052.1 | - | 7 | 969 | |||
AK315586.1 | - | 6 | 762 | |||
BC028679.2 | - | 6 | 805 | 4829354 | ||
BC048012.1 | - | 5 | 760 | |||
U59919.1 | - | 6 | 765 |