Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWC25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017748.4 | NP_060218.1 | 7, 8 | 1071 | ||
NR_073428.1 | 8, 9 | 1188 | ||||
GenBank mRNA | AK000298.1 | - | 7, 8 | 1021 | ||
AK225373.1 | - | 7, 8 | 1041 | |||
AK296115.1 | - | 8, 9 | 1153 | |||
AK316077.1 | - | 6, 7 | 1017 | |||
BC008833.2 | - | 7, 8 | 1057 | 4304656 | ||
CR749362.1 | - | 3, 4 | 254 |