Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAPG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281932.1 | NP_001268861.1 | 7, 8 | 881 | ||
NM_001281933.1 | NP_001268862.1 | 6, 7 | 828 | |||
NM_017760.6 | NP_060230.5 | 6, 7 | 828 | |||
NR_104054.1 | 8, 9 | 982 | ||||
XM_005249547.2 | XP_005249604.1 | 6, 7 | 826 | |||
XM_011516356.2 | XP_011514658.1 | 6, 7 | 826 | |||
XM_011516357.2 | XP_011514659.1 | 7, 8 | 854 | |||
XM_011516358.2 | XP_011514660.1 | 6, 7 | 826 | |||
XM_017012374.1 | XP_016867863.1 | 6, 7 | 826 | |||
XM_017012375.1 | XP_016867864.1 | 6, 7 | 826 | |||
XM_017012376.1 | XP_016867865.1 | 6, 7 | 826 | |||
XM_017012378.1 | XP_016867867.1 | 4, 5 | 445 | |||
GenBank mRNA | AK000318.1 | - | 1, 2 | 86 | ||
AK092008.1 | - | 7, 8 | 816 | |||
AK315699.1 | - | 4, 5 | 445 | |||
BC020560.1 | - | 1, 2 | 152 | 3458938 | ||
BC043404.1 | - | 6, 7 | 769 | 6045248 | ||
BX537845.1 | - | 6, 7 | 795 |