Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVA1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304762.1 | NP_001291691.1 | 3 | 705 | ||
NM_018166.2 | NP_060636.1 | 3 | 777 | |||
NR_130899.1 | 2 | 608 | ||||
XM_017001627.1 | XP_016857116.1 | 2 | 899 | |||
GenBank mRNA | AK001509.1 | - | 3 | 777 | ||
BC006241.2 | - | 2 | 489 | 3951625 | ||
BC071697.1 | - | 2 | 696 | 4371667 |