Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161575.1 | NP_001155047.1 | 6 | 1177 | ||
NM_018296.5 | NP_060766.5 | 9 | 1447 | |||
XM_005256025.2 | XP_005256082.1 | 9 | 1447 | |||
XM_005256026.2 | XP_005256083.1 | 8 | 1006 | |||
XM_005256027.2 | XP_005256084.1 | 9 | 1447 | |||
XM_005256028.1 | XP_005256085.1 | 8 | 1319 | |||
XM_011523200.1 | XP_011521502.1 | 9 | 1447 | |||
XM_011523201.1 | XP_011521503.1 | 6 | 1067 | |||
XM_011523202.2 | XP_011521504.1 | 5 | 922 | |||
XM_011523203.1 | XP_011521505.1 | 6 | 829 | |||
XM_011523204.1 | XP_011521506.1 | 7 | 1246 | |||
XM_011523205.1 | XP_011521507.1 | 8 | 1337 | |||
XM_011523206.2 | XP_011521508.1 | 9 | 1447 | |||
XM_017023400.1 | XP_016878889.1 | 9 | 1447 | |||
GenBank mRNA | AK001866.1 | - | 6 | 1226 | ||
AK292311.1 | - | 6 | 1156 | |||
AK297329.1 | - | 4 | 671 | |||
AK301766.1 | - | 6 | 1131 | |||
AK309897.1 | - | 5 | 719 | |||
AK311487.1 | - | 4 | 613 | |||
AY313780.1 | - | 6 | 1140 | |||
AY963267.1 | - | 9 | 1447 | |||
BC018031.1 | - | 8 | 1009 | |||
BC026156.2 | - | 9 | 1447 | 4828487 |