Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018467.3 | NP_060937.1 | 8 | 737 | ||
XM_017026975.1 | XP_016882464.1 | 9 | 1447 | |||
XM_017026976.1 | XP_016882465.1 | 7 | 687 | |||
GenBank mRNA | AB097050.1 | - | 8 | 708 | ||
AF220052.1 | - | 8 | 708 | |||
BC006005.1 | - | 7 | 518 | 4279653 | ||
BC008455.1 | - | 8 | 685 | 4247241 |