Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBNDD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048221.2 | NP_001041686.1 | 4 | 1126 | ||
NM_001048222.2 | NP_001041687.1 | 4 | 1224 | |||
NM_001048223.2 | NP_001041688.1 | 4 | 1075 | |||
NM_001048224.2 | NP_001041689.1 | 4 | 1173 | |||
NM_001048225.2 | NP_001041690.2 | 3 | 1401 | |||
NM_001048226.2 | NP_001041691.2 | 3 | 1499 | |||
NM_001197139.1 | NP_001184068.1 | 4 | 959 | |||
NM_001197140.1 | NP_001184069.1 | 4 | 1005 | |||
NM_018478.3 | NP_060948.3 | 4 | 1447 | |||
GenBank mRNA | AF220191.1 | - | 4 | 1453 | ||
AJ276469.1 | - | 4 | 1044 | |||
AK000531.1 | - | 4 | 957 | |||
AL591565.1 | - | 4 | 959 | |||
AY113697.1 | - | 3 | 1053 | |||
BC001105.2 | - | 4 | 969 | 2959575 | ||
BC012818.2 | - | 4 | 969 | 2959575 |
Gene Symbol: SYS1-DBNDD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003189.2 | 6 | 1310 | |||
GenBank mRNA | BC023562.2 | - | 6 | 1279 |