Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285394.1 | NP_001272323.1 | 3 | 249 | ||
NM_178817.3 | NP_848932.1 | 4 | 365 | |||
NM_206898.1 | NP_996781.1 | 4 | 365 | |||
XM_006724028.3 | XP_006724091.1 | 2 | 295 | |||
XM_017028407.1 | XP_016883896.1 | 2 | 291 | |||
GenBank mRNA | AF454915.1 | - | 4 | 365 | ||
AF454916.1 | - | 3 | 249 | |||
AF483549.1 | - | 2 | 178 | |||
AY079152.1 | - | 2 | 178 | |||
BC062721.1 | - | 2 | 303 | 30327887 |