Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HABP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014282.3 | NP_055097.2 | 2, 3 | 631 | ||
XM_005251812.3 | XP_005251869.1 | 2, 3 | 462 | |||
XM_017014484.1 | XP_016869973.1 | 2, 3 | 293 | |||
GenBank mRNA | AF241831.1 | - | 2, 3 | 581 | ||
BQ962055.1 | - | 2, 3 | 346 | |||
U77327.1 | - | 1, 2 | 164 |