Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127593.1 | NP_001121065.1 | 4 | 349 | ||
NM_001127595.1 | NP_001121067.1 | 4 | 331 | |||
NM_001127596.1 | NP_001121068.1 | 4 | 328 | |||
XM_011509293.2 | XP_011507595.2 | 3 | 238 | |||
GenBank mRNA | AK223268.1 | - | 3 | 210 | ||
AK223295.1 | - | 3 | 222 | |||
AK302126.1 | - | 3 | 561 | |||
BC017865.1 | - | 3 | 242 | 4690249 | ||
BC033678.1 | - | 3 | 217 | 5180561 | ||
BC036723.1 | - | 3 | 486 | 5206097 | ||
BI768140.1 | - | 3 | 486 | |||
DA944383.1 | - | 3 | 255 | |||
DA957430.1 | - | 4 | 347 | |||
KU178060.1 | - | 3 | 302 | |||
KU178061.1 | - | 3 | 302 | |||
M24853.1 | - | 1 | 104 | |||
S76824.1 | - | 1 | 131 | |||
X52645.1 | - | 3 | 227 |
Gene Symbol: FCGR3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB025256.1 | - | 3 | 227 | ||
AJ581669.1 | - | 3 | 227 | |||
AK316565.1 | - | 3 | 248 | |||
BC128562.1 | - | 3 | 211 | 40126686 | ||
DA670720.1 | - | 3 | 428 | |||
DA671001.1 | - | 3 | 413 | |||
DA672751.1 | - | 4 | 316 | |||
DA672763.1 | - | 3 | 468 | |||
DA673839.1 | - | 3 | 325 | |||
DC428702.1 | - | 3 | 556 | |||
M24854.1 | - | 3 | 292 | |||
X07934.1 | - | 3 | 227 | |||
X16863.1 | - | 3 | 227 |