Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SCMH1 SIRNA Assays ›
Gene Symbol:
SCMH1
Gene Name:
sex comb on midleg homolog 1 (Drosophila)
Gene Aliases:
Scml3
Chromosome Location:
Chr.1: 41027199 - 41242155 on Build GRCh38
UniGene ID:
Hs.571874
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22742
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SCMH1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001031694.2
NP_001026864.1
12
1785
NM_001172218.1
NP_001165689.1
13
1857
NM_001172219.1
NP_001165690.1
12
1648
NM_001172220.1
NP_001165691.1
11
1577
NM_001172221.1
NP_001165692.1
11
1672
NM_001172222.2
NP_001165693.1
8
1238
NM_012236.3
NP_036368.1
12
1786
NR_135092.1
12
1785
XM_006710462.2
XP_006710525.1
12
1648
XM_006710464.1
XP_006710527.1
11
1597
XM_011541032.2
XP_011539334.1
12
1586
XM_011541033.2
XP_011539335.1
14
2003
XM_011541034.1
XP_011539336.1
12
1648
XM_011541035.2
XP_011539337.1
12
1796
XM_011541036.2
XP_011539338.1
13
1859
XM_011541039.1
XP_011539341.1
10
1288
XM_011541040.2
XP_011539342.1
12
1648
XM_011541043.2
XP_011539345.1
14
2099
XM_011541044.1
XP_011539346.1
7
1215
XM_011541045.2
XP_011539347.2
13
1872
XM_017000698.1
XP_016856187.1
9
1401
XM_017000699.1
XP_016856188.1
11
1587
XM_017000700.1
XP_016856189.1
12
1797
XM_017000701.1
XP_016856190.1
10
1315
XM_017000702.1
XP_016856191.1
12
1662
XM_017000703.1
XP_016856192.1
12
1800
XM_017000704.1
XP_016856193.1
10
1315
XM_017000705.1
XP_016856194.1
11
1664
XM_017000706.1
XP_016856195.1
9
1254
XM_017000707.1
XP_016856196.1
11
1664
XM_017000708.1
XP_016856197.1
9
1237
XM_017000709.1
XP_016856198.1
11
1624
XM_017000710.1
XP_016856199.1
9
1264
XM_017000711.1
XP_016856200.1
9
1237
XM_017000712.1
XP_016856201.1
9
1228
XM_017000713.1
XP_016856202.1
9
1233
GenBank mRNA
AF149045.1
-
12
1759
AF149046.1
-
13
1830
AK023124.1
-
12
1615
AK299383.1
-
6
1083
AK301497.1
-
10
1627
BC009752.2
-
11
1727
3010273
BC021252.2
-
11
1685
3355251
BX640721.1
-
11
1563
CR457161.1
-
7
1233
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
chromatin remodeling
transcription, DNA-templated
spermatogenesis
anatomical structure morphogenesis
anterior/posterior pattern specification
gene silencing
protein sumoylation
negative regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0