Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIP2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014974.2 | NP_055789.1 | 36 | 4483 | ||
XM_005252426.3 | XP_005252483.1 | 37 | 4642 | |||
XM_005252427.3 | XP_005252484.1 | 37 | 4553 | |||
XM_005252428.4 | XP_005252485.1 | 38 | 4723 | |||
XM_005252430.2 | XP_005252487.2 | 20 | 2733 | |||
XM_011519428.2 | XP_011517730.1 | 38 | 4654 | |||
XM_011519429.2 | XP_011517731.1 | 37 | 4622 | |||
XM_011519430.2 | XP_011517732.1 | 37 | 4529 | |||
XM_011519431.2 | XP_011517733.1 | 36 | 4499 | |||
XM_011519432.1 | XP_011517734.1 | 37 | 4954 | |||
XM_011519433.2 | XP_011517735.1 | 21 | 3762 | |||
XM_017015977.1 | XP_016871466.1 | 21 | 2774 | |||
GenBank mRNA | AB023151.2 | - | 36 | 4481 | ||
AK309715.1 | - | 20 | 2495 | |||
BC048327.1 | - | 10 | 1170 | |||
BC063313.1 | - | 36 | 4483 | 30349267 |